Learn More
Biomatik Corporation Phospho-GATA1 (Ser310) Colorimetric Cell-Based ELISA Kit
Catalog No. 502291601
Encompass
Phospho-GATA1 (Ser310) Colorimetric Cell-Based ELISA Kit for detecting Phospho-GATA1 (Ser310) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P15976. Gene ID: 2623. Target Synonyms: Erythroid transcription factor; Eryf1; GATA-binding factor 1; GATA-1; GF-1; NF-E1 DNA-binding protein; GATA1; ERYF1; GF1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 502291601 Supplier Biomatik Corporation Supplier No. EKA51460
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Phospho-GATA1 (Ser310) Colorimetric Cell-Based ELISA Kit for detecting Phospho-GATA1 (Ser310) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P15976. Gene ID: 2623. Target Synonyms: Erythroid transcription factor; Eryf1; GATA-binding factor 1; GATA-1; GF-1; NF-E1 DNA-binding protein; GATA1; ERYF1; GF1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.