Learn More
Biomatik Corporation Phospho-GIRK1/KIR3.1/KCNJ3 (Ser185) Colorimetric Cell-Based ELISA Kit
Catalog No. 502291492
Encompass
Phospho-GIRK1/KIR3.1/KCNJ3 (Ser185) Colorimetric Cell-Based ELISA Kit for detecting Phospho-GIRK1/KIR3.1/KCNJ3 (Ser185) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P48549. Gene ID: 3760. Target Synonyms: G protein-activated inward rectifier potassium channel 1; IRK3; Inward rectifier K channel Kir3.1; KCNJ3; Kir3.1; Potassium channel; inwardly rectifying subfamily J member 3. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 502291492 Supplier Biomatik Corporation Supplier No. EKA51328
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Phospho-GIRK1/KIR3.1/KCNJ3 (Ser185) Colorimetric Cell-Based ELISA Kit for detecting Phospho-GIRK1/KIR3.1/KCNJ3 (Ser185) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P48549. Gene ID: 3760. Target Synonyms: G protein-activated inward rectifier potassium channel 1; IRK3; Inward rectifier K channel Kir3.1; KCNJ3; Kir3.1; Potassium channel; inwardly rectifying subfamily J member 3. Restrictions: For Research Use Only. Not for use in diagnostic procedures.