missing translation for 'onlineSavingsMsg'
Learn More

Electrostatics Materials

Numéro de catalogue. S52093
Modifier l'affichage
Click to view available options
Electrsttc Matls Set Indv 1/pk
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue.
S52093 Electrsttc Matls Set Indv 1/pk
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. S52093 Fournisseur Customer Channels Group (Ccg) Code fournisseur KT2000
Il en reste null

Comprend

Individual Kit:

  • Two clear acetate strips and two colored vinyl strips; two low-density polyethylene insulating strips with wooden handles; six coated pith balls; two squares of wool cloth; two squares of cotton cloth; two aluminum tubes and nylon monofilament

Class Kit:

  • Eight clear acetate strips; eight colored vinyl strips; eight low-density polyethylene insulating strips with wooden handles; 24 coated pith balls; eight squares of wool cloth; eight squares of cotton cloth; eight aluminum tubes and nylon monofilament

Support individual electrostatic experiments for class size up to 24 students.

Experiments with electrified objects and electrostatic induction can be performed easily with these kits. Choose between two different-sized kits — small or large. An electroscope is recommended for each group (See S43262, S52057 or S43267).

Spécifications

For Use With (Application) Experiments with electrified objects and electrostatic induction
For Use With (Equipment) Electroscope
Includes Two clear Acetate strips and two colored Vinyl strips, two LDPE insulating strips with Wooden handles, six coated pith balls, two squares of Wool cloth, two squares of Cotton cloth, two Aluminum tubes and Nylon monofilament
Product Type Electrostatics Materials Individual Kit