missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BTX™ Electroporation Cuvettes Plus™
Electroporation Cuvettes Plus™ for use in electroporation and electrofusion of bacteria, yeast, insect, plant and mammalian cells.
Supplier: BTX™ 450140
Description
- Cuvettes and safety stand protect both user and sample
- Compatible with many commercially available electroporators
- The cuvettes are molded with embedded polished aluminum electrodes, and gamma irradiated for guaranteed sterility
- BTX™ cuvettes can obtain high field strengths up to 25.0 kV/cm
- Three electrode gap sizes are available, 1 mm for bacteria and yeast, 2 mm for all cell types and 4 mm for mammalian cells
- Round cuvette caps are leak resistant and allow for quick and easy one-finger removal
- A 20-position cuvette rack is available - must be ordered separately
- BTX™ disposable cuvettes Plus require no maintenance
- Simply discard carefully after use
For 10 and 50 packs, the cuvettes are individually wrapped in a package that includes a disposable cuvette and a sterile transfer pipette, which allows for quick and easy removal of the sample after electroporation.
For bulk packages, the cuvettes are packaged in 24 bags containing 100 cuvettes per bag without transfer pipettes.
Specifications
| Sterile | |
| Single-use | |
| Polished Aluminum | |
| 2400x cuvettes | |
| 1 mm | |
| Electroporation of Bactera and Yeast |
| Electroporation Cuvette | |
| 90 μL | |
| Gray | |
| 100/Pk., 25 Pk. | |
| BTX Generators, BIORAD Gene Pulser and Micro Pulser Systems |