missing translation for 'onlineSavingsMsg'
Learn More

BTX™ Electroporation Cuvettes Plus™

Catalog No. BTX610
Change view
Click to view available options
Capacity (Metric):
90 μL
400 μL
800 μL
Width (Metric) Gap:
1 mm
2 mm
4 mm
Closure Color:
Blue
Gray
Yellow
Quantity:
10 Pk.
100/Pk., 25 Pk.
50 Pk.
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric) Width (Metric) Gap Closure Color Quantity
BTX610 90 μL 1 mm Gray 50 Pk.
BTX450134 90 μL 1 mm Gray 10 Pk.
BTX450140 90 μL 1 mm Gray 100/Pk., 25 Pk.
BTX450135 400 μL 2 mm Blue 10 Pk.
BTX450141 400 μL 2 mm Blue 100/Pk., 25 Pk.
BTX620 400 μL 2 mm Blue 50 Pk.
BTX450136 800 μL 4 mm Yellow 10 Pk.
BTX450142 800 μL 4 mm Yellow 100/Pk., 25 Pk.
BTX640 800 μL 4 mm Yellow 50 Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Catalog No. BTX610 Supplier BTX™ Supplier No. 45 0124

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Electroporation Cuvettes Plus™ for use in electroporation and electrofusion of bacteria, yeast, insect, plant and mammalian cells.

  • Cuvettes and safety stand protect both user and sample
  • Compatible with many commercially available electroporators
  • The cuvettes are molded with embedded polished aluminum electrodes, and gamma irradiated for guaranteed sterility
  • BTX™ cuvettes can obtain high field strengths up to 25.0 kV/cm
  • Three electrode gap sizes are available, 1 mm for bacteria and yeast, 2 mm for all cell types and 4 mm for mammalian cells
  • Round cuvette caps are leak resistant and allow for quick and easy one-finger removal
  • A 20-position cuvette rack is available - must be ordered separately
  • BTX™ disposable cuvettes Plus require no maintenance
    • Simply discard carefully after use

For 10 and 50 packs, the cuvettes are individually wrapped in a package that includes a disposable cuvette and a sterile transfer pipette, which allows for quick and easy removal of the sample after electroporation.

For bulk packages, the cuvettes are packaged in 24 bags containing 100 cuvettes per bag without transfer pipettes.

Specifications

Sterility Sterile
Type Electroporation Cuvette
Disposable Single-use
Capacity (Metric) 90 μL
Material Polished Aluminum
Closure Color Gray
Includes 50 x indivually wrapped cuvettes, Sterile pipet for removing the sample after electroporation
Quantity 50 Pk.
Width (Metric) Gap 1 mm
For Use With (Equipment) BTX Generators, BIORAD Gene Pulser and Micro Pulser Systems
For Use With (Application) Electroporation of Bacteria
Show More Show Less