missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Eisco™ Electrodes for Student Cell
High quality electrodes for use with cells.
$6.04 - $17.14
Specifications
| Product Type | Cell Electrode |
|---|---|
| For Use With (Application) | Student demonstration cell |
| Catalog Number | Mfr. No. | Material | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Material | Price | Quantity & Availability | |||||
|
S41554
|
Eisco™
PH0927C |
Carbon |
Each for $16.86
|
|
|||||
|
S14045
|
Eisco™
PH0927E |
Copper |
Each for $13.11
|
|
|||||
|
S14048
|
Eisco™
PH0927J |
Iron |
Each for $6.04
|
|
|||||
|
S24897
|
Eisco™
PH0927I |
Zinc |
Each for $17.14
|
|
|||||
Description
- For student demonstration cell
- Available in a wide variety of materials
Specifications
| Cell Electrode | |
| Student demonstration cell |