missing translation for 'onlineSavingsMsg'
Learn More

Electrode Storage Solution for Storing Glass and Combination pH Electrodes, Reagents

Catalog No. 01335400
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
20 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01335400 500 mL
01335403 1 L
01335402 20 L
01335401 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 01335400 Supplier Labchem Inc Supplier No. CE001600500A

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Electrode Storage Solution for Storing Glass and Combination pH Electrodes, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Specifications

Boiling Point 100°C
Color Colorless
For Use With (Equipment) Electrode
Packaging Poly Bottle
Quantity 500 mL
Solubility Completely miscible