missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Electrode Storage Solution for pH Electrodes, Reagents
Specifications
Specifications
| Boiling Point | 100°C |
| Color | Colorless |
| For Use With (Equipment) | Electrode |
| Packaging | Poly Bottle |
| Quantity | 500 mL |
| Solubility | Completely miscible |