missing translation for 'onlineSavingsMsg'
Learn More

Electrode Storage Solution, Fisher BioReagents™

Catalog No. bp24181
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BP24181 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BP24181 Supplier Fisher BioReagents Supplier No. BP24181
Only null left
Edge

For use in storage of pH electrodes

Electrode Storage Solution is a safe and convenient method to maintain sensitivity by removing buildup from electrodes.

Specifications

Chemical Name or Material Electrode Storage Solution
Color Undesignated
Melting Point 0°C
Density 1g/cm³
Synonym dihydrogen oxide, dihydrogen monoxide
Identification Pass Test
Packaging Poly Bottle
Residue after Evaporation 1.57 to 1.73%
Vapor Pressure 14mmHg at 20°C
Quantity 1 L
Physical Form Liquid
Show More Show Less