missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Electrode Cleaning Solution for Removal of Membrane/Junction Protein Deposits, Reagents
Specifications
Specifications
| Boiling Point | 100°C |
| Color | Pale Yellow |
| For Use With (Equipment) | Electrode |
| Packaging | Poly Bottle |
| Quantity | 4 L |
| Solubility | Completely miscible |