missing translation for 'onlineSavingsMsg'
Learn More

Corning™ Elastic Sleeves for CoolRack Modules

Catalog No. 07210092
Change view
Click to view available options
Quantity:
4/pk.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
07210092 4/pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 07210092 Supplier Corning™ Supplier No. 432092

For use with CoolRack Modules. Corning™ Elastic Sleeves for CoolRack Modules provides comfortable grip when handling cold, warm or wet CoolRack tube modules.

  • Colorful elastic sleeves to color-code experiments
  • Compatible with ice, dry ice and water baths
  • Four colors: purple, green, orange, pink

Sleeves fit the following modules:

  • All 15-well CoolRack modules
  • All 30-well CoolRack modules
  • CoolRack 15mL
  • CoolRack 50mL
  • CoolRack VS13
  • CoolRack V13
  • CoolRack V16

Specifications

Color Green, Orange, Purple, Pink
For Use With (Application) Color-coding CoolRack tube modules
For Use With (Equipment) CoolRack Modules
Quantity 4/pk.
Type Elastic Sleeves