Learn More
Brady™ Elastic Confined Space Cover
Get a compliant sign and barrier in one. Enhance visibility, reinforce compliance and keep unauthorized personnel out of confined spaces with a Confined Space Cover.
Supplier: Brady™ 177440
Description
Get a compliant sign and barrier in one. Enhance visibility, reinforce compliance and keep unauthorized personnel out of confined spaces with an Elastic Confined Space Cover.
- Elastic construction enables cover to wrap cover around bolts, hinges and other cover accessories or obstructions with ease.
- DANGER sign is compliant with OSHA CFR 1910.146(c)(2) and cover helps prevent unauthorized entry for two-pronged safety.
- Washable, strong material offers long-lasting durability in various industrial and environmental conditions.
- Mesh construction helps prevent entry while allowing beneficial airflow.
- Bright colors and reflective elements enhance visibility.
- Compact and lightweight for easy transport and storage.
Chemical Identifiers
| OSHA 1910.146 | |
| Round | |
| DANGER | |
| Red |
| Polyester | |
| English | |
| Confined Space Sign |
Specifications
| OSHA 1910.146 | |
| Fits standard 30 in. opening | |
| Round | |
| DANGER | |
| Red |
| Polyester | |
| 30 in. | |
| English | |
| Confined Space Sign |