missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Elastatinal, Thermo Scientific Chemicals
Irreversible elastase inhibitor
$268.33 - $268.33
Chemical Identifiers
| CAS | 51798-45-9 |
|---|---|
| Molecular Formula | C21H36N8O7 |
| Molecular Weight (g/mol) | 512.568 |
| MDL Number | MFCD00151180 |
| InChI Key | IJWCGVPEDDQUDE-WXZHEDMLSA-N |
| Synonym | elastatinal, leu-co-cpd-gln-ala-h |
| PubChem CID | 15942859 |
| IUPAC Name | (2S)-2-[[(1S)-2-[[(2S)-5-amino-1,5-dioxo-1-[[(2S)-1-oxopropan-2-yl]amino]pentan-2-yl]amino]-1-(2-amino-1,4,5,6-tetrahydropyrimidin-6-yl)-2-oxoethyl]carbamoylamino]-4-methylpentanoic acid |
| SMILES | CC(C)CC(C(=O)O)NC(=O)NC(C1CCN=C(N1)N)C(=O)NC(CCC(=O)N)C(=O)NC(C)C=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAJ61120LB0
|
Thermo Scientific Chemicals
J61120LB0 |
5 mg |
Each for $268.33
|
|
|||||
Description
Irreversible elastase inhibitor
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 51798-45-9 | |
| 512.568 | |
| IJWCGVPEDDQUDE-WXZHEDMLSA-N | |
| 15942859 | |
| CC(C)CC(C(=O)O)NC(=O)NC(C1CCN=C(N1)N)C(=O)NC(CCC(=O)N)C(=O)NC(C)C=O |
| C21H36N8O7 | |
| MFCD00151180 | |
| elastatinal, leu-co-cpd-gln-ala-h | |
| (2S)-2-[[(1S)-2-[[(2S)-5-amino-1,5-dioxo-1-[[(2S)-1-oxopropan-2-yl]amino]pentan-2-yl]amino]-1-(2-amino-1,4,5,6-tetrahydropyrimidin-6-yl)-2-oxoethyl]carbamoylamino]-4-methylpentanoic acid |
Specifications
| 51798-45-9 | |
| C21H36N8O7 | |
| 5 mg | |
| elastatinal, leu-co-cpd-gln-ala-h | |
| CC(C)CC(C(=O)O)NC(=O)NC(C1CCN=C(N1)N)C(=O)NC(CCC(=O)N)C(=O)NC(C)C=O | |
| 512.568 | |
| 512.6 | |
| Elastatinal |
| White to Yellow | |
| MFCD00151180 | |
| 873543 | |
| IJWCGVPEDDQUDE-WXZHEDMLSA-N | |
| (2S)-2-[[(1S)-2-[[(2S)-5-amino-1,5-dioxo-1-[[(2S)-1-oxopropan-2-yl]amino]pentan-2-yl]amino]-1-(2-amino-1,4,5,6-tetrahydropyrimidin-6-yl)-2-oxoethyl]carbamoylamino]-4-methylpentanoic acid | |
| 15942859 | |
| Powder |
Safety and Handling
EINECSNumber : 257-426-7
RTECSNumber : LZ9600000
TSCA : No
Recommended Storage : Store at -20°C
RUO – Research Use Only