Two times crystallized, (Code: ESL), supplied as a dialyzed, lyophilized powder. The enzyme should be 0.22 micron filtered after reconstitution and prior to use. Suitable for the isolation of Type II lung cells. Store at 2-8C.
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Two times crystallized, (Code: ESL), supplied as a dialyzed, lyophilized powder. The enzyme should be 0.22 micron filtered after reconstitution and prior to use. Suitable for the isolation of Type II lung cells. Store at 2-8C.