missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Elaidic Acid 97.0+%, TCI America™
$225.56 - $820.19
Chemical Identifiers
| CAS | 112-79-8 |
|---|---|
| Molecular Formula | C18H34O2 |
| Molecular Weight (g/mol) | 282.468 |
| MDL Number | MFCD00063954 |
| InChI Key | ZQPPMHVWECSIRJ-MDZDMXLPSA-N |
| Synonym | elaidic acid, trans-oleic acid, trans-9-octadecenoic acid, 9-octadecenoic acid, e-oleic acid, 9-octadecenoic acid, e, e-octadec-9-enoic acid, trans-elaidic acid, 9-octadecenoic acid, 9e, elaidinsaure |
| PubChem CID | 637517 |
| ChEBI | CHEBI:27997 |
| IUPAC Name | (E)-octadec-9-enoic acid |
| SMILES | CCCCCCCCC=CCCCCCCCC(=O)O |
Chemical Identifiers
| 112-79-8 | |
| 282.468 | |
| ZQPPMHVWECSIRJ-MDZDMXLPSA-N | |
| 637517 | |
| (E)-octadec-9-enoic acid |
| C18H34O2 | |
| MFCD00063954 | |
| elaidic acid, trans-oleic acid, trans-9-octadecenoic acid, 9-octadecenoic acid, e-oleic acid, 9-octadecenoic acid, e, e-octadec-9-enoic acid, trans-elaidic acid, 9-octadecenoic acid, 9e, elaidinsaure | |
| CHEBI:27997 | |
| CCCCCCCCC=CCCCCCCCC(=O)O |
Specifications
| 112-79-8 | |
| White-Yellow | |
| C18H34O2 | |
| 5 g | |
| ZQPPMHVWECSIRJ-MDZDMXLPSA-N | |
| (E)-octadec-9-enoic acid | |
| 637517 | |
| 282.47 | |
| Crystalline Powder |
| 44°C | |
| 288°C | |
| MFCD00063954 | |
| elaidic acid, trans-oleic acid, trans-9-octadecenoic acid, 9-octadecenoic acid, e-oleic acid, 9-octadecenoic acid, e, e-octadec-9-enoic acid, trans-elaidic acid, 9-octadecenoic acid, 9e, elaidinsaure | |
| CCCCCCCCC=CCCCCCCCC(=O)O | |
| 282.468 | |
| CHEBI:27997 | |
| ≥97.0% (T) | |
| Elaidic Acid |
Safety and Handling
EINECSNumber : (2)-0609
RTECSNumber : JX6125000
TSCA : No
Recommended Storage : Refrigerator