missing translation for 'onlineSavingsMsg'
Learn More

Elaidic acid, 98%

Store cold | CAS: 112-79-8 | C18H34O2 | 282.468 g/mol

Supplier:  Thermo Scientific Chemicals A1483206

 View more versions of this product

Catalog No. AAA1483206


Only null left
Explore available promotions

Description

Description

In human platelets incubated with arachidonic acid, elaidic acid inhibits HHT and HETE formation while inducing prostaglandin and thromboxane synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
In human platelets incubated with arachidonic acid, elaidic acid inhibits HHT and HETE formation while inducing prostaglandin and thromboxane synthesis.

Solubility
Insoluble in water.

Notes
Air Sensitive. Keep container tightly closed. Store away from oxidizing agents.
Specifications

Specifications

Elaidic acid
41°C to 45°C
>110°C (230°F)
C18H34O2
5 g
14,3533
Insoluble in water.
CCCCCCCCC=CCCCCCCCC(=O)O
282.468
CHEBI:27997
98%
112-79-8
288°C (100 mmHg)
98%
MFCD00063954
1726543
elaidic acid, trans-oleic acid, trans-9-octadecenoic acid, 9-octadecenoic acid, e-oleic acid, 9-octadecenoic acid, e, e-octadec-9-enoic acid, trans-elaidic acid, 9-octadecenoic acid, 9e, elaidinsaure
ZQPPMHVWECSIRJ-MDZDMXLPSA-N
(E)-octadec-9-enoic acid
637517
282.47

Chemical Identifiers

112-79-8
282.468
ZQPPMHVWECSIRJ-MDZDMXLPSA-N
637517
(E)-octadec-9-enoic acid
C18H34O2
MFCD00063954
elaidic acid, trans-oleic acid, trans-9-octadecenoic acid, 9-octadecenoic acid, e-oleic acid, 9-octadecenoic acid, e, e-octadec-9-enoic acid, trans-elaidic acid, 9-octadecenoic acid, 9e, elaidinsaure
CHEBI:27997
CCCCCCCCC=CCCCCCCCC(=O)O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 204-006-6

RTECSNumber : JX6125000

TSCA : No

storageNote1 : Air sensitive

Recommended Storage : Keep cold; Store under Nitrogen

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only