missing translation for 'onlineSavingsMsg'
Learn More

Elacridar

Catalog No. AC464952500
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC464952500 250 mg
AC464950010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC464952500 Supplier Thermo Scientific Chemicals Supplier No. 464952500
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 143664-11-3 | C34H33N3O5 | 563.65 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 143664-11-3
Molecular Formula C34H33N3O5
Molecular Weight (g/mol) 563.65
InChI Key OSFCMRGOZNQUSW-UHFFFAOYSA-N
IUPAC Name N-{4-[2-(6,7-dimethoxy-1,2,3,4-tetrahydroisoquinolin-2-yl)ethyl]phenyl}-5-methoxy-9-oxo-9,10-dihydroacridine-4-carboxamide
SMILES COC1=C2NC3=C(C=CC=C3C(=O)C2=CC=C1)C(=O)NC1=CC=C(CCN2CCC3=CC(OC)=C(OC)C=C3C2)C=C1

Specifications

Melting Point 210.0°C
Quantity 250 mg
Formula Weight 563.65
Chemical Name or Material Elacridar

RUO – Research Use Only