Learn More
Eisco™ Visual Scientifics Wave Demonstration Set
Catalog No. S07814
Change view
Click to view available options
Wave Demonstration set
Catalog No.
S07814 Wave Demonstration set
1 options
Catalog No. S07814 Supplier Eisco™ Supplier No. PTWDA
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Includes
Thin and thick strings, oscillating motor, mountable pulley
Requires
Visual Scientific's base (S06893) and back board (S06894), ring stand posts, video camera, digital balance
Guides students through understanding standing waves, nodes, tension and speed.
  • Helps students understand various concepts like: effect of amplitude of pulse, length, tension and material of spring on speed of pulse
  • Includes 10+ page instruction booklet covering approximately two to three hours of class/lab time material, 10+ Next Generation Science Standards Concepts and 10+ Common Core Standards
  • Every module includes comprehensive instructions to assist with lesson planning, student activities and experimental procedures
  • All labs and extensions feature recommended video analysis setups and guides
Specifications
Product Type Wave Demonstration Set
For Use With (Application) Guides students through understanding standing waves, nodes, tension and speed
Includes Thin and thick strings, oscillating motor, mountable pulley, instructions, student activities, experimental procedures