missing translation for 'onlineSavingsMsg'
Learn More

Eisco™ Visual Scientifics Physical Science System Base

Catalog No. S06893
Change view
Click to view available options
Grade:
9 to 12
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S06893 9 to 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S06893 Supplier Eisco™ Supplier No. PTBASE

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Visual Scientifics Physical Science System Base is a system that allows students to use all technologies available to learn physics through technology, engineering and real-world applications.

A large, robust base with precise magnetic and threaded mounting locations helps students set up experiments easily and quickly with reproducible results.

  • Large, magnetic base for sturdily attaching experiments
  • High-contrast color grid for easy data collection with video analysis systems
  • Each junction in the grid accepts threaded rods for accurate mounting
  • Dimensions: 25 x 16 x 2.5 in. (63.5 x 40.6 x 6.35 cm)
  • Accepts any probeware system

Order Info

For use with S06894, S06895, S06896, S06897, S06898, S06899, S06900, S06901, S06902, S06903.

Specifications

Grade 9 to 12
Description Magnetic base
Format Kit
Product Type Physical Science System Base
Includes Laser, Slide Holder, Diffraction Grating (500 lines/mm), Diffraction Grating (1000 lines/mm), (2) Visual Scientifics Posts, Laboratory manual and student worksheet
Width (English) 6 in.
Width (Metric) 152.4 mm
Quantity 1
Color Black
Languages English
For Use With (Application) A system that allows students to use all technologies available to learn physics through technology, engineering and real-world applications
Subject Physics
Show More Show Less