Learn More
Eisco™ Visual Scientifics Diffraction Experiment Set
The Visual Scientifics Diffraction Experiment Set helps students derive and understand diffraction and the key building blocks of modern physics.
Supplier: Eisco™ PTYDSE
Includes: Mounted laser, Visual Scientifics posts, slide holder, diffraction grating 1, diffraction grating 2, diffraction grating 3
Requires : Ruler
Description
- Experimentally prove y=mλL/d and understand more about wave and particle duality.
- Fast and easy to set up and explore.
- Also comes with a 10+ page instruction booklet covering approximately two to three hours of class/lab time material, 10+ Next Generation Science Standards Concepts, and 10+ Common Core Standards.
- Every module includes comprehensive instructions to assist with lesson planning, student activities and experimental procedures.
- All labs and extensions feature recommended video analysis setups and guides.
Specifications
| Helps students derive and understand diffraction and the key building blocks of modern physics | |
| Mounted Laser, Visual Scientifics Posts, Slide Holder, Diffraction Grating 1, Diffraction Grating 2, Diffraction Grating 3, Instructions Student Activities, Experimental Procedures | |
| 1 Ea. | |
| Visual Scientifics Diffraction Experiment Set |