Learn More
Eisco™ Solar System Model
Catalog No. S05842
Change view
Click to view available options
Quantity:
Each
Catalog No. Quantity
S05842 Each
1 options
Catalog No. S05842 Supplier Eisco™ Supplier No. BD0074A
Three dimensional model.
  • Students can see the planets in their relatives sizes and colours. Planet may be set by month to show their relative position
  • Sturdy base supports 15 cm. dia. Sun with the outer planet 18 in. from the Sun
  • Illustrated study guide includes planet position tables
Specifications
For Use With (Application) Students can see the planets in their relatives sizes and colors
Diameter (Metric) 15 cm
Diameter (English) 5.9 in.
Product Type 3D Solar System Model
Quantity Each