missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Eisco™ Secchi Disk Including Rope
Description
Includes
Instruction manual
- Has nine in. disc of alternating black and white quadrants
- Provides quick and easy way to probe turbidity, which is measure of clarity of water
- Turbidity is in turn related to amount of sediments or pollutants in water
Specifications
Specifications
| Grade | 9 to College |
| Product Type | Secchi Disk |
| Includes | Secchi Disk, 10+ Ft. of String, Small Handle |
| For Use With (Application) | Quick qualitative measurements of the clarity of water over time |