missing translation for 'onlineSavingsMsg'
Learn More

Eisco™ Pulley Model

Catalog No. S07820
Change view
Click to view available options
Pulley Model
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
S07820 Pulley Model
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S07820 Supplier Eisco™ Supplier No. WDMS12

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Includes

Activity guide

Demonstrates how the use of several pulleys can increase mechanical advantage.

  • Three pulleys mounted on a solid wood base
  • Sturdy wooden construction is ideal for hands-on classroom use
  • Can be used both qualitatively and quantitatively to study mechanical advantage
  • Simple design lends itself easily to inquiry-based activities

Specifications

Product Type Pulley Model
Material Wood
For Use With (Application) Demonstrate how use of several pulleys increases mechanical advantage
Includes Activity guide