missing translation for 'onlineSavingsMsg'
Learn More

Eisco™ Pulley Demonstration Kit

Catalog No. S02104
Change view
Click to view available options
Class Size:
1 to 2 Lab Groups
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S02104 1 to 2 Lab Groups
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S02104 Supplier Eisco™ Supplier No. PLPSES

Includes

  • Rail (14.3 in. [36.3cm])
  • Two aluminum rods Two clamp sliders Two boss heads Five slotted masses (three 50g, two 100g)
  • Weight holder
  • Pulley with rod
  • Pulley on hook
  • Instructions

Requires

NeuLog™ Force Logger Sensor (S98243-9ND)

Use the Pulley Demonstration Kit to investigate one of the six simple machines.

Observe how pulleys can change the direction of an applied force, transmit rotational motion or carry out a mechanical advantage in either linear or rotational system of motion.

Kit enables students to study:

  • General properties of pulleys
  • Pulley systems

Specifications

Grade 7 to Undergraduate
For Use With (Application) Used to observe how pulleys can change the direction of an applied force, transmit rotational motion or carry out a mechanical advantage in either linear or rotational system of motion
Includes 14.3 in. Rail, 2 Aluminum Rods, 2 Clamp Sliders, 2 Boss Heads, 5 Slotted Masses (3 x 50 G, 2 x 100 G), Weight Holder, Pulley with Rod, Pulley on Hook, Instructions
Class Size 1 to 2 Lab Groups
Product Type Pulley Demonstration Kit