Learn More
Eisco™ Physics Kit: Electrostatics
To understand fundamental concepts of static electricity with the Electrostatics Physics Kit.
Supplier:  Eisco™ EIRQ04
Catalog No. S13467
Only null left
Explore available promotions
Description
Description
  • Over 25 components
  • Use to teach different activities
  • Includes Activity Guides with Student worksheets
Activities Covered
  • Exploring Electrostatic Charge Using Rods and Cloths
  • Movement of Charge on an Object
  • Is There More Than One Type of Charge
  • Comparing Magnetic and Electrostatic Forces
  • The Electroscope
  • Conductors versus Insulators
  • Charging objects by Induction and Conduction
  • Volta's Electrophorus
  • Coulomb's Law
  • The Conducting Cup
  • Faraday's Ice Pail
Includes:
Wool Cloth 26 x 26 cm (1), Silk Cloth (1), Glass rod (1), Ebonite rod (1), Perspex rod (1), Wire stirrup (1), Polyethylene strip (1), Gold leaf electroscope (1), Gold leaf (1), Glass panes that go on either side of the electroscope (2), Metal conducting knob to collect charge on top of the electroscope (1), Metal conducting plate to collect charge on top of the electroscope (1), Metal conducting bucket (Faraday Pail) to collect charge of top of the electroscope (1), Pith balls connected by string (1), Pith ball electroscope stand (1), Base for electroscope or metal conductive spheres 3, Metal conducting spheres with insulating post (one with hole in the top) (2), Electrophorus disk with handle (1), Square polyethylene tile (1), Neon tube (1), Reel of nylon filament Diameter 0.20 mm, spool of 25 m (1), Small electrophorus disk with handle (1)
Specifications
Specifications
7 to Undergraduate
1 Wool Cloth 26 x 26 cm, 1 Silk Cloth, 1 Glass Rod, 1 Ebonite Rod, 1 Perspex Rod, 1 Wire Stirrup, 1 Polyethylene Strip, 1 Gold leaf Elecroscope, 1 Gold leaf, 2 Glass panes, 1 Metal Conducting knob, 1 Metal Conducting plate, 1 Metal Conducting Bucket (Faraday Pail), 1 Pith Balls Connected By String, 1 Pith Ball Elecroscope Stand, 3 Bases, 2 Metal Conducting Spheres, 1 Electrophorus Disk with Handle
1 Lab Group
Electrostatics Kit
Product Suggestions
Product Suggestions