Learn More
Eisco™ Magdeburg Hemisphere Rubber
Description
Student place the edges of two hemisphere cups together and squeeze them, forcing out the air between them. In a tug of war, students try to pull the cups apart, surprisingly difficult task.
- With two-finger tug handle and two release posts
- 7.5cm diameter
Specifications
Specifications
| Product Type | Magdeburg Hemisphere |
| Material | Rubber |
| For Use With (Application) | Demonstrates the force of air pressure without a vacuum pump |
| Includes | 2 finger tug handle and 2 release posts |