missing translation for 'onlineSavingsMsg'
Learn More

Eisco™ Lab Essential Kit- Superior

Catalog No. S02632
Change view
Click to view available options
Class Size:
25 Students
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S02632 25 Students
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S02632 Supplier Eisco™ Supplier No. LABKIT
Get Flask Sets, Goggles, or a Kit with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Includes

One each of base 20 x 12.5cm and 600mm rod, beaker tong, crucible tong, tripod stand, wire gauge, bosshead, clamp, retort ring with boss head, burette clamp double, test tube holder, test tube stand Z shape, test tube brush and spatula.

Required for use in chemistry lab experiments.

Kit Components (one each of all items below):

  • Base
  • Rod
  • Beaker tong
  • Crucible tong
  • Tripod stand
  • Wire gauge
  • Boss head
  • Clamp
  • Retort ring with boss head
  • Burette clamp double
  • Test tube holder
  • Test tube stand (z-shape)
  • Test tube brush - spatula

Specifications

Grade 9 to 12
Includes One each of base 20 x 12.5 cm and 600 mm rod, beaker tong, crucible tong, tripod stand, wire gauge, bosshead, clamp, retort ring with boss head, burette clamp double, test tube holder, test tube stand z shape, test tube brush and spatula
Class Size 25 Students
Product Type Superior Lab Essential Kit