missing translation for 'onlineSavingsMsg'
Learn More

Eisco™ Giant Human Heart Model

Catalog No. S15621
Change view
Click to view available options
Dimensions (L x W x H):
10.0 x 9.0 x 8.0 in.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W x H)
S15621 10.0 x 9.0 x 8.0 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S15621 Supplier Eisco™ Supplier No. AM75AS

Get an amazing look at the human heart with this 3 times life size model.

Dissectible into 7 parts, it shows excellent detail of both internal and external structures. Great for study of the human circulation and the movement of blood through the hearts. Sits on a 9 x 8 in. base.

Specifications

Dimensions (L x W x H) 10.0 x 9.0 x 8.0 in.
Includes Numbered with English Key Card; calls out 23 structures
Material ABS Plastic
Mounting Base
Size 3x Life-Size
Weight (English) 4.8 lb.
Product Type Heart Model