missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Eisco™ Gear Train Model
Description
Includes
Activity guide
- Three identical wooden gears linearly mounted with hand crank
- Demonstrates how gears operate at a basic level to make work easier
- Enables students to visualize concepts of work, force and energy
Specifications
Specifications
| Product Type | Model, Gear Train |
| Material | Wood |
| For Use With (Application) | Demonstrate how gears work |
| Includes | Activity guide |