missing translation for 'onlineSavingsMsg'
Learn More

Eisco™ Gear Rack Model

Catalog No. S07823
Change view
Click to view available options
Gear Rack Model
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
S07823 Gear Rack Model
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S07823 Supplier Eisco™ Supplier No. WDMS15

Includes

Activity guide

Simple gear and rack system demonstrates how gears turn rotational motion into translational motion.

  • Enables students to visualize how machines can reduce work
  • Sturdy wooden construction is ideal for hands-on classroom use

Specifications

Product Type Model, Gear Rack
Material Wood
For Use With (Application) Classroom use, demonstrates how gears can turn rotational motion into translational motion
Includes Activity guide