missing translation for 'onlineSavingsMsg'
Learn More

Eisco™ Electrostatic Friction Rod Kit

Catalog No. S13000
Change view
Click to view available options
Class Size:
3 Lab Groups
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S13000 3 Lab Groups
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S13000 Supplier Eisco™ Supplier No. PH1120150

Use the Electrostatic Friction Rod Kit to study static electricity by creating charges with different materials.

  • This electrostatic kit provides the necessary tools to free both positive and negative charges for exploration into electrostatic forces.
  • Students can experiment with different combinations of cloth and rod materials to explore how each manufactures charge carriers.

Specifications

Grade 6 to 11
For Use With (Application) Demonstrations of both positive and negative charge
Includes Wool Cloth, Cotton Cloth, Silk Cloth, Glass Rod, Ebonite Rod, Lucite Rod
Class Size 3 Lab Groups
Product Type Electrostatic Friction Rod Kit