Learn More
Eisco™ Electroscope Kit
Catalog No. S12981
Change view
Click to view available options
Electroscope Kit
Catalog No.
S12981 Electroscope Kit
1 options
Catalog No. S12981 Supplier Eisco™ Supplier No. PH0901
Includes
Pair of flask type electroscopes, two each discs and terminals, one Faraday's ice pail, spare foil leaf pairs and instructions.
For electroscope experiments and other related studies.
For experiments of electroscope and other related studies such as electrostatic induction, the nature of charge in a body, Faraday’s ice pail experiment etc.
Specifications
For Use With (Application) For experiments of electroscope and other related studies
Includes Pair of flask type electroscope, two each discs and terminals, one Faraday's ice pail, spare foil leaf pairs and instructions.
Product Type Electroscope Kit