missing translation for 'onlineSavingsMsg'
Learn More

Eisco™ Drilled Ball Set

Catalog No. S07846
Change view
Click to view available options
Class Size:
1 Lab Group
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S07846 1 Lab Group
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S07846 Supplier Eisco™ Supplier No. PH0109CB

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Classroom Drilled Ball Set enables learning of pendulums, density and mass experiments.

  • 12 balls made of aluminum, brass, cork, lead, steel and wood.
  • Enclosed in plastic case.

Specifications

Grade 7 to 12
For Use With (Application) Used for learning pendulums, density and mass experiments
Includes 12 Balls of Aluminum, Brass, Cork, Lead, Steel, and Wood
Material Aluminum, Brass, Cork, Lead, Steel, Wood
Class Size 1 Lab Group
Product Type Drilled Ball Set