Learn More
Eisco™ Direct Vision Spectroscope
Catalog No. S99345
Change view
Click to view available options
Direct vision spectroscope
Catalog No.
S99345 Direct vision spectroscope
1 options
Catalog No. S99345 Supplier Eisco™ Supplier No. PH0594
For rapid examination of spectral composition of white light.
  • Spectroscope consists of a metal tube with draw-out focusing and an adjustable slit
  • Slit fitted with achromatic objective glass and three element prisms
  • Comes in a wooden case
Specifications
For Use With (Application) Rapid examination of spectral composition of white light
Includes Metal tube with draw out focusing , adjustable slit at 1 end and an eyepiece mounted on a draw tube at the other end fitted with achromatic objective glass, 3 element prisms, Wooden case
Product Type Direct Vision Spectroscope