missing translation for 'onlineSavingsMsg'
Learn More

Eisco™ Density Block Set with Hooks

Catalog No. S07847
Change view
Click to view available options
Product Type:
Density Block Set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
S07847 Density Block Set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S07847 Supplier Eisco™ Supplier No. PH0108PCB

Perfect for studying density.

  • Consists of four 32mm cubes made of aluminum, brass, lead and steel with hooks
  • Attached hooks makes handling of material easy

Specifications

For Use With (Application) To study density
Product Type Density Block Set
Includes Set of four cube Brass, Lead, Aluminum, Steel with hook