missing translation for 'onlineSavingsMsg'
Learn More

Eisco™ Boss Head Thermometer Holder

Catalog No. S95951
Change view
Click to view available options
Includes:
Hook
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes
S95951 Hook
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S95951 Supplier Eisco™ Supplier No. CH0667

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Great way to suspend a thermometer.

  • Die cast with metal alloy finish
  • Hook on one end for suspending thermometer

Specifications

Disposable Reusable
For Use With (Application) Suspend a thermometer over a hot plate or a specific heat specimen
Includes Hook
Product Type Suspension Clamp
Description Holds items 110 mm from support rod
For Use With (Equipment) Support Rod, Thermometer
Color Gray