Learn More
Eisco™ Bimetallic Strip Demonstration
Catalog No. S23666
Change view
Click to view available options
Grade:
7 to 12
Catalog No. Grade
S23666 7 to 12
1 options
Catalog No. S23666 Supplier Eisco™ Supplier No. PH0383
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Demonstrates the principles of thermal expansion in metals. Bimetallic Strip Demonstration allows students to understand how bimetallic strips are applied practically in things such as thermometers, thermostats and heating coils.
  • Hands on demonstration of expansion differences between various metals.
  • Single piece non-jointed.
  • Wooden handle for safe experimentation.
  • Strip comprised of copper and steel metals.
  • Includes activity guide.
Specifications
Grade 7 to 12
Includes 1 Strip comprised of Copper and Steel attached to Wood Handle
Size 12.2 x 7.5 x 4.3 in.
Dimensions (L x W x H) 12.2 x 7.5 x 4.3 in.
Product Type Bimetallic Strip Demonstration