missing translation for 'onlineSavingsMsg'
Learn More

Eisco™ Bar Magnets

Catalog No. S95965
Change view
Click to view available options
Color:
Red and Blue
Znc-plated steel
Shape:
Bar
Rectangular
Weight (English):
0.95 lb.
3.5 lb.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Shape Weight (English)
S95965 Red and Blue Bar 3.5 lb.
S95966 Znc-plated steel Rectangular 0.95 lb.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options

CHOKING HAZARD (1). Small parts. Not for children under 3 yrs.

Catalog No. S95965 Supplier Eisco™ Supplier No. PH0786D

CHOKING HAZARD (1). Small parts. Not for children under 3 yrs.

Perfect for classroom experiments.

  • Red and blue magnets with stamped polarity measure 3.9 x 0.79 x 0.23 in. (10 x 2 x 0.6cm) and are boxed
  • Rectangular square-faced chrome steel magnets measure 5.9 x 0.4 x 0.4 in. (15 x 1 x 1cm)

Order Info

Magnets sold in packs of two.

Specifications

Material Chrome Steel
Shape Bar
For Use With (Application) Perfect for classroom experiments
Grade 9 to 12
Color Red and Blue
Product Type Bar Magnet
Includes Keeper
Weight (Metric) 1.59 kg
Width (English) 5.51 in.
Weight (English) 3.5 lb.
Width (Metric) 14 cm
Show More Show Less