missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Eicosanoic acid, 98%

CAS: 506-30-9 | C20H40O2 | 312.54 g/mol

$63.55 - $287.77

Chemical Identifiers

CAS 506-30-9
Molecular Formula C20H40O2
Molecular Weight (g/mol) 312.54
MDL Number MFCD00002755
InChI Key VKOBVWXKNCXXDE-UHFFFAOYSA-N
Synonym arachidic acid, eicosanoic acid, arachic acid, n-eicosanoic acid, arachidinic acid, arachidate, eicosanoate, unii-pqb8mjd4rb, pqb8mjd4rb, n-eicosanoate
PubChem CID 10467
ChEBI CHEBI:28822
IUPAC Name icosanoic acid
SMILES CCCCCCCCCCCCCCCCCCCC(O)=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC117720010
SDS View Documents
Thermo Scientific Chemicals
117720010
1 g Glass bottle
Each for $63.55
Only null left
 
AC117720100
SDS View Documents
Thermo Scientific Chemicals
117720100
10 g Glass bottle
Each for $287.77
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

506-30-9
312.54
VKOBVWXKNCXXDE-UHFFFAOYSA-N
10467
icosanoic acid
C20H40O2
MFCD00002755
arachidic acid, eicosanoic acid, arachic acid, n-eicosanoic acid, arachidinic acid, arachidate, eicosanoate, unii-pqb8mjd4rb, pqb8mjd4rb, n-eicosanoate
CHEBI:28822
CCCCCCCCCCCCCCCCCCCC(O)=O

Specifications

506-30-9
White
Authentic
Glass bottle
CH3(CH2)18CO2H
1 g
arachidic acid, eicosanoic acid, arachic acid, n-eicosanoic acid, arachidinic acid, arachidate, eicosanoate, unii-pqb8mjd4rb, pqb8mjd4rb, n-eicosanoate
VKOBVWXKNCXXDE-UHFFFAOYSA-N
icosanoic acid
10467
312.52
Glistening Powder or Flakes
74.0°C to 77.0°C
328.0°C
99%
C20H40O2
MFCD00002755
15, 753
Solubility in water: practically insoluble. Other solubilities: freely soluble in hot absolute alcohol, benzene,, chloroform and ether
CCCCCCCCCCCCCCCCCCCC(O)=O
312.54
CHEBI:28822
99%
Eicosanoic acid
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 208-031-3

SDS
Documents

Documents

RUO – Research Use Only