missing translation for 'onlineSavingsMsg'
Learn More

Eicosanedioic Acid 98.0+%, TCI America™

Catalog No. p-7128933
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E032025G 25 g
E0320250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E032025G Supplier TCI America Supplier No. E032025G
Only null left

Chemical Identifiers

CAS 2424-92-2
Molecular Formula C20H38O4
Molecular Weight (g/mol) 342.52
MDL Number MFCD00059627
InChI Key JJOJFIHJIRWASH-UHFFFAOYSA-N
Synonym eicosanedioic acid, unii-sod235tgnz, 1,18-octadecanedicarboxylic acid, sod235tgnz, octadecane-1,18-dicarboxylic acid, eicosanedioicacid, 1,20-icosanedioic acid, 1,20-eicosanedioic acid, eicosa-1,20-dioic acid
PubChem CID 75502
ChEBI CHEBI:73728
IUPAC Name icosanedioic acid
SMILES C(CCCCCCCCCC(=O)O)CCCCCCCCC(=O)O

Specifications

Melting Point 127°C
Color White
Boiling Point 234°C
Quantity 25 g
Formula Weight 342.52
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Eicosanedioic Acid
EINECSNumber (2)-3083
TSCA Yes