missing translation for 'onlineSavingsMsg'
Learn More

Eicosane 98.0+%, TCI America™

Catalog No. E000325G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E000325G 25 g
E0003100G 100 g
E0003500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. E000325G Supplier TCI America Supplier No. E000325G

Chemical Identifiers

CAS 112-95-8
Molecular Formula C20H42
Molecular Weight (g/mol) 282.56
MDL Number MFCD00009344
InChI Key CBFCDTFDPHXCNY-UHFFFAOYSA-N
Synonym eicosane, n-eicosane, unii-3aya9kec48, ccris 663, nonadecane, methyl, 3aya9kec48, eicosane, analytical standard, octyldodecane, n-icosane
PubChem CID 8222
ChEBI CHEBI:43619
IUPAC Name icosane
SMILES CCCCCCCCCCCCCCCCCCCC

Specifications

Melting Point 36°C
Color White
Boiling Point 344°C
Quantity 25 g
Formula Weight 282.56
Percent Purity ≥98.0% (GC)
Physical Form Crystals
Chemical Name or Material Eicosane
EINECSNumber (2)-0010
TSCA Yes