missing translation for 'onlineSavingsMsg'
Learn More

Eicosane 99.5+%, TCI America™

Catalog No. S02925ML
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S02925ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S02925ML Supplier TCI America Supplier No. S02925ML

Chemical Identifiers

CAS 112-95-8
Molecular Formula C20H42
Molecular Weight (g/mol) 282.56
MDL Number MFCD00009344
InChI Key CBFCDTFDPHXCNY-UHFFFAOYSA-N
Synonym eicosane, n-eicosane, unii-3aya9kec48, ccris 663, nonadecane, methyl, 3aya9kec48, eicosane, analytical standard, octyldodecane, n-icosane
PubChem CID 8222
ChEBI CHEBI:43619
IUPAC Name icosane
SMILES CCCCCCCCCCCCCCCCCCCC

Specifications

Melting Point 36°C
Color White
Boiling Point 344°C
Quantity 5 mL
Formula Weight 282.56
Percent Purity ≥99.5% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material Eicosane [Standard Material for GC]
EINECSNumber (2)-0010
TSCA Yes