Learn More
Lonza Walkersville Inc EGM -2 MV Microvascular Endothelial Cell Growth Medium-2 BulletKit
Supplier:  Lonza Walkersville Inc CC3202
Encompass
Choose from a variety of endothelial cell growth media for an easy cultivation of primary endothelial cells. Every endothelial cell growth medium has its special formulation and is optimized to grow certain cell types. Each type of endothelial cell growth medium is guaranteed to perform when used to grow Clonetics Primary Cells.Content:1 x EBM-2 Basal Medium (CC-3156),500 mL1 x EGM-2 MV SingleQuots Supplement Pack (CC-4147) containing:1 x Bottle FBS, 25.00 mL1 x Natural Cap Vial with Hydrocortisone, 0.20 mL1 x Gray Cap Vial with hFGF-B, 2.00 mL1 x White Cap Vial with VEGF, 0.50 mL1 x Yellow Cap Vial with R3-IGF-1, 0.50 mL1 x Blue Cap Vial with Ascorbic Acid, 0.50 mL1 x Green Cap Vial with hEGF, 0.50 mL1 x Red Cap Vial with GA-1000, 0.50 m
Catalog No. NC9902887
Only null left