missing translation for 'onlineSavingsMsg'
Learn More

Ferric Subsulfate Solution, USP, 20-22 g/100 mL, Spectrum™ Chemical

Catalog No. 18602682
Change view
Click to view available options
Quantity:
500 mL
6 x 500 mL
200 L
Packaging:
Amber Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18602682 200 L Amber Glass Bottle
18602680 500 mL Amber Glass Bottle
18602681 6 x 500 mL Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18602682 Supplier Spectrum Chemical Mfg Cor Supplier No. F1305200LTBL
Get Up to $300 in Additional Chemicals
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

F1305, 1310-45-8, Fe4(OH)2(SO4)5

Spectrum™ Chemical Ferric Subsulfate Solution, USP All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 1310-45-8
Molecular Formula Fe4H2O22S5
Molecular Weight (g/mol) 737.67
InChI Key GDPKWKCLDUOTMP-UHFFFAOYSA-B
IUPAC Name tetrairon(3+) dihydroxide pentasulfate
SMILES [OH-].[OH-].[Fe+3].[Fe+3].[Fe+3].[Fe+3].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O

Specifications

CAS Max % 100%
Physical Form Liquid
Quantity 200 L
Grade USP
Concentration or Composition (by Analyte or Components) 20 to 22 g/100 mL
Packaging Amber Glass Bottle
Chemical Name or Material Ferric Subsulfate Solution