missing translation for 'onlineSavingsMsg'
Learn More

Efavirenz 98.0+%, TCI America™

Catalog No. E09975G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E09975G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. E09975G Supplier TCI America Supplier No. E09975G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 154598-52-4
Molecular Formula C14H9ClF3NO2
Molecular Weight (g/mol) 315.676
MDL Number MFCD05662344
InChI Key XPOQHMRABVBWPR-ZDUSSCGKSA-N
Synonym (4S)-6-Chloro-4-(2-cyclopropylethynyl)-1,4-dihydro-4-(trifluoromethyl)-2H-3,1-benzoxazin-2-one
PubChem CID 64139
ChEBI CHEBI:119486
IUPAC Name (4S)-6-chloro-4-(2-cyclopropylethynyl)-4-(trifluoromethyl)-1H-3,1-benzoxazin-2-one
SMILES C1CC1C#CC2(C3=C(C=CC(=C3)Cl)NC(=O)O2)C(F)(F)F

Specifications

Melting Point 138°C
Color White-Yellow
Quantity 5 g
Formula Weight 315.68
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material Efavirenz
RTECSNumber DM3440000
TSCA No
Recommended Storage Freezer