Learn More
Edwards & Cromwell Universal Leak Control, AE Series Kits
Catalog No. 19811089
Encompass
Change view
Click to view available options
For Use With (Equipment):
Non-sparking tools
Catalog No. For Use With (Equipment)
19811089 Non-sparking tools
1 options
Catalog No. 19811089 Supplier Edwards & Cromwell Supplier No. AENS
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Kit contains commonly used components for emergency and temporary leak stoppage of liquid or bulk hazardous materials due to punctures, gashes, cracks, or surface rotting.
  • Designed for small to medium holes in any type of low-pressure vessel
  • Covers pin, convex, concave, and flat surface holes as well as cracks of all shapes
  • Employs quick application, twin “T” patch and ladder patch design for average forklift-type punctures
These kits designed for use only by experienced and trained personnel only for temporary control until the container can be disposed of properly. Proper protective clothing must be worn.
Specifications
For Use With (Equipment) Non-sparking tools