missing translation for 'onlineSavingsMsg'
Learn More

Edvotek™ M12 Complete™ Electrophoresis Package

Catalog No. S90242ND
Change view
Click to view available options
Capacity:
2 Gel Trays
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity
S90242ND 2 Gel Trays
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S90242ND Supplier Edvotek™ Supplier No. EMD504

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Includes

(2) 7 x 7 cm Gel Trays, (1) 7 x 14 cm Gel Tray, (2) 6 Tooth Combs, (2) 8/10 Tooth Combs, (4) Rubber End Caps

Student-friendly horizontal electrophoresis apparatus for DNA, dye, and protein agarose gels. Includes trays, combs, and end caps.

Run the full spectrum of horizontal electrophoresis experiments with this versatile package! The reimagined M12 Complete™ supports one or two student groups in two standard length gel trays for experiments that require less separation, or one long gel tray for experiments that require more. Produces excellent results in 30-40 minutes and includes a lifetime warranty.

Specifications

Capacity 2 Gel Trays
Detection Method Electrophoresis
For Use With (Application) Gel Electrophoresis
Gel Size Mini
Max. Current 500 mA
Running Time 15 to 30 min.
Timer 0 to 99 hr. or continuous
Type Horizontal Electrophoresis System
Includes One 7 x 14cm trays, two 6-teeth combs, one 8/10-teeth comb, four Rubber end caps
Model M12 dual
Sample Size Up to 20
Volume (English) Buffer 13.5 oz.
Volume (Metric) Buffer 400 mL
Warranty Lifetime
Length (English) Gel 2.7 in.
Length (Metric) Gel 7 cm
Width (English) Gel 2.7 in.
Width (Metric) Gel 7 cm
Max. Voltage 200 V
Frequency 50/60 Hz
Show More Show Less