missing translation for 'onlineSavingsMsg'
Learn More

EDTA Disodium Salt, Ultrapure, 500 g, Innovating Science™

Catalog No. S000968
Encompass
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S000968 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S000968 Supplier Aldon Corporation Supplier No. IS29039
  • Appearance: White Crystalline Powder
  • Purity: ≥ 99.0%
  • pH (5% Aqueous): 4.0 to 6.0
  • Solubility: Clear and Colorless to Faint Yellow
  • Identification (FTIR): Conforms to Structure
  • Loss on Drying (4h at 105 °C): ≤ 1.0%
  • Insoluble Matter: ≤ 0.005%
  • Nitriloacetic Acid: ≤ 0.1%
  • Lead: ≤ 5 ppm
  • Iron: ≤ 10 ppm
  • DNase: None Detected
  • RNase: None Detected

Chemical Identifiers

Molecular Weight (g/mol) 372.24

Specifications

Quantity 500 g
Percent Purity 100%
Grade Ultra Pure
Physical Form Crystalline Powder or Crystals