missing translation for 'onlineSavingsMsg'
Learn More

EDTA, Disodium Salt, DILUT-IT™ Analytical Concentrate, 0.1M, 1/10 mole = 33.62g C10H14N2Na2O8, J.T. Baker™

Catalog No. 02002781
Change view
Click to view available options
Quantity:
1 Pk.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
02002781 1 Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 02002781 Supplier Avantor Supplier No. 465301

Chemical Identifiers

CAS 6381-92-6
Molecular Formula C10H18N2Na2O10
Molecular Weight (g/mol) 372.24
MDL Number MFCD00150037,MFCD00003541
InChI Key OVBJJZOQPCKUOR-UHFFFAOYSA-L
Synonym edta, edetic acid, ethylenediaminetetraacetic acid, edathamil, versene, endrate, havidote, titriplex, edta acid, sequestrol
PubChem CID 6049
ChEBI CHEBI:42191
IUPAC Name disodium 2-({2-[(carboxylatomethyl)(carboxymethyl)amino]ethyl}(carboxymethyl)amino)acetate dihydrate
SMILES O.O.[Na+].[Na+].OC(=O)CN(CCN(CC(O)=O)CC([O-])=O)CC([O-])=O

Specifications

Quantity 1 Pk.
Chemical Name or Material EDTA, Disodium Salt, DILUT-IT™ Analytical Concentrate, 0.1M, 1/10 mole = 33.62g C10H14N2Na2O8