missing translation for 'onlineSavingsMsg'
Learn More

EDTA, Disodium Salt (Dihydrate), Molecular Biology Grade, Promega™ DFS Item

A chelator of divalent metal cations.

Supplier:  Promega Corporation H5032

Catalog No. PRH5032

Only null left
Explore available promotions

Description

Description

  • Quality tested and certified free of DNase, RNase and protease activity
Specifications

Specifications

Ethylenediaminetetraacetic acid, disodium salt
(0.1M in water), 0.2 max. at 260nm
1g/cu.cm
Authentic
C10H14N2Na2O8
500 g
0.005% max. Insoluble Matter
[Na+].[Na+].OC(=O)CN(CCN(CC(O)=O)CC([O-])=O)CC([O-])=O
336.21
CHEBI:64734
≥99.0%
Crystalline Powder
139-33-3
White
4 to 5 (5% soln. at 25°C)
≥99+%
MFCD00070672,MFCD00003541,MFCD00070672,MFCD00150037
ethylenediaminetetraacetic acid disodium salt, edta na2, disodium 2-2-bis carboxymethyl amino ethyl-carboxymethyl amino acetic acid
ZGTMUACCHSMWAC-UHFFFAOYSA-L
disodium 2-({2-[(carboxylatomethyl)(carboxymethyl)amino]ethyl}(carboxymethyl)amino)acetate
57339238
372.2
Molecular Biology
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if inhaled.

GHS P Statement:
IF INHALED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

TSCA : TSCA

Recommended Storage : 15 to 30°C

SDS
Documents

Documents

Promotions

Promotions